pH Combination electrodes
Sensolyt® EC*, Sensolyt® EC, without arming, for the installation into flow vessels, 2 - 12, Normal contaminated water
Codice catalogo WTWO109102
Nome del fornitore: WTW ONLINE
Vai a
Product Details & Documents
The SensoLyt® measuring chains with arming are easy to install and boast flexible measuring and application areas. Eletrodes without arming can be connected directly to a pH monitor of the EcoLine® or QuadroLine® series. Depending on the application, they can be installed into a flow vessel.
- Ideal for measurements in water and drinking water
- Also for high pressure and temperature conditions
- Easy to maintain by easy measuring chain replacement
Specifiche
- Per:Normal contaminated water
- Tipo:Sensolyt® EC, without arming, for the installation into flow vessels
- Descrizione:Sensolyt® EC*
- Seq:32
- pH:2 - 12