Orders LinkContactUsLinkComponent
Stopcocks, Rotaflo®
3 mm, 10 mm
  BIBBHP3RA/7
 :   SciLabware
Stopcocks, Rotaflo®
  BIBBHP3RA/7
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
3 mm, 10 mm
SFr. 154.00
1 * 1 ST
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
  • Ø foro:
    3 mm
  • Seq:
    15
  • Per tubo con Ø interno:
    10 mm
Accessories
  •  
    Descrizione
    Per
  • 229-0230
    Replacement key, bore Ø 6 mm
    High performance Rotaflo® stopcocks
    1 * 2 ST - SFr. 161.00
  •  
    Descrizione
    Per
  • 229-0231
    Replacement key, bore Ø 6 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - SFr. 222.00
  •  
    Descrizione
    Per
  • BIBBLR6
    Replacement locking ring for key with bore 6 mm
    Rotaflo® stopcocks
    1 * 5 ST - SFr. 53.00
  •  
    Descrizione
    Per
  • 229-0226
    Replacement key, bore Ø 10 mm
    High performance Rotaflo® stopcocks
    1 * 1 ST - SFr. 103.00
  •  
    Descrizione
    Per
  • 229-0229
    Replacement key, bore Ø 3 mm
    High performance Rotaflo® stopcocks
    1 * 2 ST - SFr. 115.00
  •  
    Descrizione
    Per
  • BIBBLR3
    Replacement locking ring for key with bore 3 mm
    Rotaflo® stopcocks
    1 * 5 ST - SFr. 36.10
  •  
    Descrizione
    Per
  • 229-0234
    Replacement key, bore Ø 10 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - SFr. 345.00
  •  
    Descrizione
    Per
  • BIBBGP3K
    Replacement key, bore Ø 3 mm
    General purpose Rotaflo® stopcocks
    1 * 5 ST - SFr. 142.00
  •  
    Descrizione
    Per
  • BIBBLR10
    Replacement locking ring for key with bore 10 mm
    Rotaflo® stopcocks
    1 * 5 ST - SFr. 49.50