Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Trays, large deep

Trays, large deep

bianco, 50 l, 550×430×310 mm
Catalog # DELT19603
Supplier:   DELTALAB
Trays, large deep
Catalog # DELT19603
Supplier:  DELTALAB
Trays, large deep

bianco, 50 l, 550×430×310 mm

Price
SFr. 80.50
Log in for availability
1 * 1 ST
Product Details & Documents

Trays made of natural HDPE.

  • Stackable
  • Good temperature resistance
  • Good chemical resistance

With ergonomically designed handles.

Specifications
  • Capacità:
    50 l
  • Colore:
    bianco
  • Seq:
    01
  • L×P×A:
    550×430×310 mm