Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Powder funnels, PP/PE

Powder funnels, PP/PE

80 mm, 21 mm
Codice catalogo BURK9606-0080
Nome del fornitore:   BURKLE
Powder funnels, PP/PE
Codice catalogo BURK9606-0080
Nome del fornitore:  BURKLE
Powder funnels, PP/PE
Powder funnels, PP/PE

80 mm, 21 mm

Price
SFr. 4.60
Log in per disponibilità
1 * 1 ST
Product Details & Documents

These powder funnels are designed for safe filling of containers.

  • PP/PE

9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).

Specifiche
  • Ø esterno imbuto:
    80 mm
  • Seq:
    01
  • Ø esterno stelo:
    21 mm