Orders LinkContactUsLinkComponent
Condensers, Graham
Condensers, Graham
400 mm, 24/29 NS, - NS
  WITG1.733.400
Condensers, Graham
  WITG1.733.400
Condensers, Graham
Condensers, Graham
400 mm, 24/29 NS, - NS
SFr. 161.00
1 * 1 ST
Borosilicate glass 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Coil type, Graham pattern, suitable for either reflux or distillation work. Water connection with glass olives.
  • Dimensione cono:
    24/29 NS
  • Lunghezza mantello:
    400 mm
  • Cono femmina:
    - NS