Quality control test suspensions
Pseudomonas aeruginosa, 2,5 mL
Codice catalogo NAMSGP25-07
Vai a
Discontinued
Product Details & Documents
The control suspension (growth promotion) contains <100 CFU`s/0,1 ml.
- 2,5 ml pro vial
- 25 dose
Specifiche
Specifiche
- Conf:2,5 mL
- Descrizione:Pseudomonas aeruginosa