Orders LinkContactUsLinkComponent
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
EV-100-R, 100 µl, -, -, Trasparente, MCA96 / HV96, 50 rack da 96
  736-0447
 :   Corning
Automation tips for Tecan®, Axygen®
  736-0447
 :  Corning
Automation tips for Tecan®, Axygen®
Automation tips for Tecan®, Axygen®
EV-100-R, 100 µl, -, -, Trasparente, MCA96 / HV96, 50 rack da 96
SFr. 689.00
1 * 4.800 ST
Polipropilenedisponibili, con filtro e senza filtro, sterili e non sterili
  • Puntali neri conduttivi per protocolli di rilevamento del livello di liquido
  • Punta fine ottimizzata per un'erogazione accurata
  • Certificati esenti da RNasi, DNasi ed endotossine
These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
EVO-style tips are compatible with Tecan Freedom EVO®, Caliper Zephyr® SPE, and BioTek® Precision™ robotic pipetting systems.
Genesis-style tips are compatible with Tecan Genesis Freedom, Freedom EVO, and MiniPrep robotic workstations, as well as Cavro®, PerkinElmer/Packard Multiprobe II HT, and Multiprobe II HT EX workstations.
Confezione: Packaged in plastic blister packs.
Attenzione: * Confezione economica ad avvolgimento in cartone
  • Confezione:
    50 rack da 96
  • Sterili:
    -
  • Volume:
    100 µl
  • Seq:
    28
  • Stile punta:
    EV-100-R
  • Colore puntale:
    Trasparente
  • Per uso con:
    MCA96 / HV96
  • Filtro:
    -
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..