D-(-)-Lyxose ≥99%
Glass bottle for solids, 25 g
Purity: 99% CAS No.: 1114-34-7 Molecular Formula: C5H10O5 Formula Weight: 150.13 Melting Point: 109-115° Beilstein Reference: 1723083 Merck Reference: 14,5641 MDL No.: MFCD00064362
- :1114-34-7
- :C₅H₁₀O₅
- :MFCD00064362
- :150.13 g/mol
- :14,5641
- :109…115 °C
- :1,545
- :Ambiente
- :214-212-8
- Tipo di conf.:Glass bottle for solids
- Conf:25 g