Orders LinkContactUsLinkComponent
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® - 1 tube with 1,37 ml of SRK® solution in 10 ml tube + 1 plastic applicator attached to the cap, viscose tip, in bulk; Rayon wrapped regular tip; 50×6, bulk; 16×100 skirted sferic bottom tube, rosso
Codice catalogo 710-0932
Nome del fornitore:   Copan
SRK® Surfaces and Equipment Microbial Testing Devices
Codice catalogo 710-0932
Nome del fornitore:  Copan
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® Surfaces and Equipment Microbial Testing Devices
SRK® - 1 tube with 1,37 ml of SRK® solution in 10 ml tube + 1 plastic applicator attached to the cap, viscose tip, in bulk; Rayon wrapped regular tip; 50×6, bulk; 16×100 skirted sferic bottom tube, rosso
Price
SFr. 279.00
Log in per disponibilità
1 * 300 ST
Product Details & Documents
Our Swab Rinse Kits line includes comprehensive tailor-made collection and devices for the detection of environmental microbial contamination in a broad range of environments, such as food production, pharmaceutical sites, and hospital wards. Inspectors and Quality Assurance personnel can use SRK® products for routine surface monitoring within manufacturing areas, hospital surfaces, and equipment.
  • Optimal recovery rate: Choose the sampling probe that best suits your needs: regular FLOQSwabs® and CLASSIQSwabs™, plastic/viscose applicator, or sponge spatula
    Compliance: SRK® kits are designed compliant with ISO 18593:2018, ISO 18593:2004, ISO 17604:2003, and EU directive 2001/471/E.C
    Media choice for enhanced preservation: Choose between an isotonic solution or a wide range of selective and enrichment broths: Half Fraiser, RVS, BPW 1%, TSB, UNS, and Letheen broth
    Sterile sampling frames: the definition of a set sampling area is essential for microbial bioburden validations
    Sampling frames will make your surface sampling match that standard
This product line groups a variety of product codes, each optimised to address specific needs.
Specifiche
  • Descrizione:
    SRK® - 1 tube with 1,37 ml of SRK® solution in 10 ml tube + 1 plastic applicator attached to the cap, viscose tip, in bulk; Rayon wrapped regular tip; 50×6, bulk; 16×100 skirted sferic bottom tube
  • Seq:
    18
  • Colore tappo:
    rosso