Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
 

Accessories for test sieves

Receiver for beechwood frame test sieves, 300 mm, 80 mm
  HAVE205922794
 :   NEXOPART
Accessories for test sieves
  HAVE205922794
 :  NEXOPART
Accessories for test sieves
Accessories for test sieves

Receiver for beechwood frame test sieves, 300 mm, 80 mm

 
102.00 SFr.
   
1 * 1 ST
 
 
  • Ø:
    300 mm
  • Descrizione:
    Receiver for beechwood frame test sieves
  • Altezza:
    80 mm
  • Seq:
    12
  • Cat. No.:
    HAVE205922794
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.