Pipette tips, ratiolab®
Makro, 1000 - 5000 µl, -, Trasparente, Pipette Gilson® Pipetman®/Neo, 50/rack
RATI2410630
: RATIOLAB
ratiolab® pipette tips are ideal for use with all commercially available pipettes.
- Made of high quality PP
- Tight fit due to the precisely fitting cone
- Guaranteed dust-free due to special ionization treatment
- Excellent transparency due to high quality surfaces
- Minimal residual wetting due to controlled quality of raw materials
- Excellent volume accuracy due to precise tip orifice
- MultiTray® refill system is space- and cost-saving and clearly reduces the amount of waste
MultiTray®: Dust-proof refill pack for the Multibox® plus and the Multibox® ultra.
- Confezione:50/rack
- Sterili:-
- Volume:1000 - 5000 µl
- Seq:37
- Stile punta:Makro
- Colore puntale:Trasparente
- Per uso con:Pipette Gilson® Pipetman®/Neo