Microtubes
single, for use with racks, 0,65 ml
Codice catalogo RATI8200021
Nome del fornitore: RATIOLAB
Vai a
Product Details & Documents
Translucent polypropylene (PP) with very low protein binding.
- Very good temperature resistance
- Very good chemical resistance
The 0.65 and 1.2 ml micro-tubes can be supplied as single, strips of 8 or in 96-well format as a refill system for 8 x 12 plates.
Specifiche
- Capacità:0,65 ml
- Descrizione:single, for use with racks