Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 15/16 mm Ø tube, 12
Codice catalogo JENC357-132
Nome del fornitore: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Codice catalogo JENC357-132
Nome del fornitore: JENCONS SCIENTIFIC
Vai a
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifiche
- N. di fori:12
- Descrizione:Interchangeable block
- Seq:11
- Per:15/16 mm Ø tube