Interchangeable blocks for block heaters, digital, SB1
Interchangeable plain block, without holes, -, —
Codice catalogo JENC357-122
Nome del fornitore: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Codice catalogo JENC357-122
Nome del fornitore: JENCONS SCIENTIFIC
Vai a
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifiche
- N. di fori:—
- Descrizione:Interchangeable plain block, without holes
- Seq:01
- Per:-