Pycnometer
5 ml
Codice catalogo HECH42571008
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in stopper
- Good chemical resistance
- Good temperature resistance
Certifications: DIN ISO 3507
Specifiche
- Volume (nom.):5 ml