Cylinders, without lid
80 mm
Codice catalogo HECH44409018
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Jars, made of glass or stainless steel, with base, without lids.
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.
Ordering information: Lids with knob available must be ordered separately.
Specifiche
- Ø est:80 mm
- Seq:17