Funnels with short stem
160 mm
Codice catalogo HECH49050018
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean
Specifiche
- Ø esterno imbuto:160 mm