Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
140 mm
Codice catalogo 221-0316
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Funnels with short stem
Codice catalogo 221-0316
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Funnels with short stem
Funnels with short stem
140 mm
Price
SFr. 4.30
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
  • Good chemical resistance
  • Suspension eye for easy storage
  • Easy to clean
Specifiche
  • Ø esterno imbuto:
    140 mm