Funnels with short stem
120 mm
Codice catalogo HECH49050012
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean
Specifiche
- Ø esterno imbuto:120 mm