Orders LinkContactUsLinkComponent
Funnels with short stem
60 mm
  HECH49050003
Funnels with short stem
  HECH49050003
Funnels with short stem
Funnels with short stem
60 mm
SFr. 2.00
1 * 1 ST
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
  • Good chemical resistance
  • Suspension eye for easy storage
  • Easy to clean
  • Ø esterno imbuto:
    60 mm