Funnels with short stem
60 mm
HECH49050003
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean
- Ø esterno imbuto:60 mm