Cork borer sets
Cork borers, set of 12, 5,0 - 19,0 mm
Codice catalogo HECH41593012
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
These cork borers are made of nickel-plated brass tubes with handles.
Specifiche
- Ø foro:5,0 - 19,0 mm
- Descrizione:Cork borers, set of 12
- Seq:02