Orders LinkContactUsLinkComponent
Pressure bulbs
Rubber bulbs, single, with valve and tubing
Codice catalogo HECH40850001
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Pressure bulbs
Codice catalogo HECH40850001
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Pressure bulbs
Pressure bulbs
Rubber bulbs, single, with valve and tubing
Price
SFr. 14.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents
simple: with 2 valves – made of soft PVC (cadmium-free)
double: with valve - made of soft PVC (cadmium-free), ball in the net made of latex
Specifiche
  • Descrizione:
    Rubber bulbs, single, with valve and tubing
  • Seq:
    01