Flasks, flat bottom with standard ground joint
500 ml, 29/32 NS
Catalog # HECH41911024
Supplier: GLASWARENFABRIK KARL HECHT
Flasks, flat bottom with standard ground joint
Catalog # HECH41911024
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Specifications
- Cono femmina:29/32 NS
- Capacità:500 ml
- Seq:05