Flasks, flat bottom with standard ground joint
50 ml, 29/32 NS
Codice catalogo HECH41911006
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Flasks, flat bottom with standard ground joint
Codice catalogo HECH41911006
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Specifiche
- Cono femmina:29/32 NS
- Capacità:50 ml
- Seq:01