Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
50 ml, 29/32 NS
Codice catalogo HECH41911006
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Flasks, flat bottom with standard ground joint
Codice catalogo HECH41911006
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
Flasks, flat bottom with standard ground joint
Flasks, flat bottom with standard ground joint
50 ml, 29/32 NS
Price
16.80 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Specifiche
  • Cono femmina:
    29/32 NS
  • Capacità:
    50 ml
  • Seq:
    01