Wire baskets, square
200×200×200 mm
Codice catalogo HECH41075020
Nome del fornitore: GLASWARENFABRIK KARL HECHT
Vai a
Product Details & Documents
These square baskets are made of stainless steel. Designed for transporting, storing, cleaning and sterilising laboratory utensils.
- Excellent temperature resistance
- Good chemical resistance
- Reinforced edges
Specifiche
- L×P×A:200×200×200 mm