Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Scintillation vials, with snap cap

Scintillation vials, with snap cap

20,00 ml, 26×60 mm, PP, Yes, Snap cap
Codice catalogo SEMA3825
Nome del fornitore:   SEMADENI
Scintillation vials, with snap cap
Codice catalogo SEMA3825
Nome del fornitore:  SEMADENI
Scintillation vials, with snap cap
Scintillation vials, with snap cap

20,00 ml, 26×60 mm, PP, Yes, Snap cap

Price
61.00 SFr.
Log in per disponibilità
1 * 100 ST
Product Details & Documents

These scintillation vials are made of high quality material and good solvent resistance with good stability.

  • With patented quick-release fastener
  • Closures are easily removable
Specifiche
  • Tappo incluso:
    Yes
  • Ø×A:
    26×60 mm
  • Capacità:
    20,00 ml
  • Materiale:
    PP
  • Seq:
    02
  • Tipologia tappo:
    Snap cap