Notepads, microplate wells
Tampone pozzetto 96 per micropiastre, 127×178 mm
Codice catalogo 613-1455
Nome del fornitore: DIVERSIFIED BIOTECH
Vai a
Product Details & Documents
These pads with the layout of a 24-, 48- or 96-well microtiter plate provide a fast and easy way to keep accurate records of samples in a microplate. The adhesive backing allows the researcher to affix individual sheets into a notebook.
- With adhesive backing for attaching individual sheets
- Encourages accurate record keeping
Supplied with 50 pads of 127×178 mm.
Specifiche
- Descrizione:Tampone pozzetto 96 per micropiastre, 127×178 mm
- Seq:01