L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 500 ml
Codice catalogo SH30034.02
Nome del fornitore: HyClone products (Cytiva)
L-Glutamine, HyClone™
Codice catalogo SH30034.02
Nome del fornitore: HyClone products (Cytiva)
Numero CAS:
Vai a
Product Details & Documents
HyClone™ L-glutamine is used as a supplement in serum- and protein-free media. It supports the growth of cells that have high energy demands and synthesise large amounts of proteins and nucleic acids.
Documents
Certificates
Cerca per numero di lottoGE Healthcare Cell Boost™ 7a and 7b Supplements
Product Literature
Specifiche
Specifiche
- Conf:500 ml
- Descrizione:L-Glutamine 200 mM solution, concentration 29,2 mg/ml