Orders LinkContactUsLinkComponent
L-Glutamine, HyClone™
L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 500 ml
Codice catalogo SH30034.02
Nome del fornitore:   HyClone products (Cytiva)
L-Glutamine, HyClone™
Codice catalogo SH30034.02
Nome del fornitore:  HyClone products (Cytiva)
Numero CAS:  
L-Glutamine, HyClone™
L-Glutamine 200 mM solution, concentration 29,2 mg/ml, 500 ml
Numero CAS:  
Price
34.60 SFr.
Log in per disponibilità
1 * 500 mL
Product Details & Documents
HyClone™ L-glutamine is used as a supplement in serum- and protein-free media. It supports the growth of cells that have high energy demands and synthesise large amounts of proteins and nucleic acids.
Specifiche
Specifiche
  • Conf:
    500 ml
  • Descrizione:
    L-Glutamine 200 mM solution, concentration 29,2 mg/ml
Specifiche