Labels, hazard warning
Hazard labels: hot surface, —, Poliestere, nero on giallo, 100×87 mm, 3 labels/card
Codice catalogo 116-0683
Nome del fornitore: Brady
Vai a
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifiche
- Colore:nero on giallo
- Descrizione:Hazard labels: hot surface
- Dimensioni:100×87 mm
- Materiale:Poliestere
- Seq:27
- Modello:—
- Confezione:3 labels/card



