Sovraocchiali, 2800
Anti-UV, antigraffio, antiappannamento, blu, In PC trasparente, 2C - 1,2
Codice catalogo 111-1603
Nome del fornitore: 3M
Vai a
Product Details & Documents
These lightweight cover glasses (OTG) are designed to fit over most prescription spectacles with minimal interference. Available in three lens options. The green lens offers protection against infrared radiation arising from certain gas welding, gas cutting and brazing applications (IR shade 5).
- Strong polycarbonate lens with anti-scratch coating
- Adjustable temple length (four positions)
- Inclinable lens for easy adjustment and maximum comfort
- Soft low profile temples for comfort and minimum interaction with prescription spectacles
Weight: 38 g
: EN 166, EN 170, EN 171 (IR), FT1
Specifiche
- Tipo:Anti-UV, antigraffio, antiappannamento
- Colore:blu
- Materiale lente:In PC trasparente, 2C - 1,2