Microplates, 96-well
Microplate, 96-well, U-bottom, without lid, trasparente, 96
Codice catalogo BOET05.011.0100
Nome del fornitore: BOETTGER PAUL
Vai a
Product Details & Documents
PS, optically clear, without lids. Microplates with a choice of different well types for different applications. The plates are fully compatible with standard reading and washing equipment.
- Safe stacking, no danger of slipping
- Gripping surfaces at the side of the plate
- Frosted surface prevents errors from extraneous light sources
- Alphanumerical well identification for easy sample location
- Marking area on the side
- Lids (optional) have condensation rings to reduce evaporation
- Lid orientation corners reduce risk of cross-contamination
Specifiche
- N. di pozzetti:96
- Colore:trasparente
- Descrizione:Microplate, 96-well, U-bottom, without lid
- Seq:02