Plugs for test tubes
LDPE, 16 mm, naturale
Codice catalogo 212-0518
Nome del fornitore: BOETTGER PAUL
Vai a
Product Details & Documents
Polyethylene (PE). Ribbed plugs with two sealing ribs, very tight sealing.
- Not autoclavable
- Easy handling with a high, ribbed grip
- Easy to open by tilting 'one-hand operation'
Specifiche
- Colore:naturale
- Materiale:LDPE
- Seq:06
- Taglia:16 mm