Insoles, Image
47, verde
Codice catalogo 114-0497
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
Colour: light green.
- Non woven fibres, honeycomb structure, hypoallergenic
- Impregnated with anti-fungal, antibacterial agent, neutralises perspiration
- Combats the main germs responsible for mycoses and other infection risks
Specifiche
- Taglia scarpe:47
- Colore:verde