Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Soluzioni ISA

Soluzioni ISA

Sulfide anti-oxidant buffer (SAOB) for sulfide ion selective electrode, 475 ml bottles, Ion selective electrode reagents, Bottle, 1 KIT
Codice catalogo 662-0833
Nome del fornitore:   Thermo Orion
Soluzioni ISA
Codice catalogo 662-0833
Nome del fornitore:  Thermo Orion
Numero CAS:  
Soluzioni ISA
Soluzioni ISA

Sulfide anti-oxidant buffer (SAOB) for sulfide ion selective electrode, 475 ml bottles, Ion selective electrode reagents, Bottle, 1 KIT

Numero CAS:  
Price
SFr. 401.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents

Ionic strength adjustors are used for analysis with ion selective electrodes, and special reagents are used with some ion selective electrodes.

  • Ionic strength adjuster is added to all samples and standards to create a uniform background ionic strength, which provides more reproducible measurements
  • Ammonia pH-adjusting ISA and low-level ammonia pH-adjusting ISA both include a pH-indicating blue dye for instant verification of correct pH in ammonia samples and standards with no pH measurement required
  • Ammonia pH-adjusting ISA prevents complexation of ammonia and inhibits formation of metal hydroxides, allowing determination of total ammonia in the sample and increasing the accuracy of results
  • Low-level ammonia pH-adjusting ISA is ideal for ammonia samples with a concentration of 10 ppm and below that do not contain metal interferences

Concentration : 10 ppm

For Use With (Equipment) : Ion selective electrode
For Use With (Application) : Ionic strength adjustors used for analysis with ion selective electrodes and special reagents used with some ion selective electrodes

Specifiche

Specifiche

  • Tipo di conf.:
    Bottle
  • Conf:
    1 KIT
  • Descrizione:
    Sulfide anti-oxidant buffer (SAOB) for sulfide ion selective electrode, 475 ml bottles
  • Tipo:
    Ion selective electrode reagents

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..