Slide Boxes
viola, PS, 50, 172×83×31 mm
Catalog # GLWAK50V
Supplier: GLW
Jump to:
Product Details & Documents
High quality slide storage boxes for microscopy slides.
Used for storing, transporting and freezing in science, industry and biotechnology.
Specifications
- N. di vetrini:50
- Colore:viola
- Materiale:PS
- Seq:07
- L×P×A:172×83×31 mm
Accessories
Product Information
- Catalog #DescrizioneColoreMaterialeAvailabilityPrice
- GLWAZK20Divider for slides and coverslips of special sizesgrigioPSLog in for availability1 * 1 ST - SFr. 3.10
Product Information
- Catalog #DescrizioneColoreMaterialeAvailabilityPrice
- GLWAZK15Divider for slides and coverslips of special sizesbiancoPSLog in for availability1 * 1 Un. - SFr. 6.50