Orders LinkContactUsLinkComponent
Insoles, Image
Insoles, Image
46, verde
Catalog # 114-0496
Supplier:   HONEYWELL SAFETY
Insoles, Image
Catalog # 114-0496
Supplier:  HONEYWELL SAFETY
Insoles, Image
Insoles, Image
46, verde
Price
SFr. 21.60
Log in for availability
1 * 1 PAIRE
Product Details & Documents
Colour: light green.
  • Non woven fibres, honeycomb structure, hypoallergenic
  • Impregnated with anti-fungal, antibacterial agent, neutralises perspiration
  • Combats the main germs responsible for mycoses and other infection risks
Specifications
  • Taglia scarpe:
    46
  • Colore:
    verde