Orders LinkContactUsLinkComponent
Insoles, Image
40, verde
Codice catalogo 114-0490
Nome del fornitore:   HONEYWELL SAFETY
Insoles, Image
Codice catalogo 114-0490
Nome del fornitore:  HONEYWELL SAFETY
Insoles, Image
Insoles, Image
40, verde
Price
21.60 SFr.
Log in per disponibilità
1 * 1 PAIRE
Product Details & Documents
Colour: light green.
  • Non woven fibres, honeycomb structure, hypoallergenic
  • Impregnated with anti-fungal, antibacterial agent, neutralises perspiration
  • Combats the main germs responsible for mycoses and other infection risks
Specifiche
  • Taglia scarpe:
    40
  • Colore:
    verde