Pipette tips
Universal, 100 - 1000 µl, -, Blu, Kartell, Eppendorf, Gilson, Socorex, Finnpipette, Brand, Rack
Codice catalogo KART9615
Nome del fornitore: KARTELL
Vai a
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- Tips in racks come with snap-on lid
- Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
- Containers and tips are autoclavable to +121 °C for 20 minutes
Specifiche
- Confezione:Rack
- Sterili:-
- Volume:100 - 1000 µl
- Seq:34
- Stile punta:Universal
- Colore puntale:Blu
- Per uso con:Kartell, Eppendorf, Gilson, Socorex, Finnpipette, Brand