Organic reference standards, ethanol FS
Ethanol standard FS, 3,0 mg/ml, Reference standard, 10×1 ml, Ampoules, 10 mL
DIAS109309910349
- Concentrazione:3,0 mg/ml
- Tipo di conf.:Ampoules
- Conf:10 mL
- Standard:Reference standard
- Descrizione:Ethanol standard FS
- Pk Info:10×1 ml
- Cat. No.:DIAS109309910349
- Formula:C₂H₅OH
- Descrizione:Ethanol
- CAS No.:64-17-5
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



