Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Leather gloves, Precision Tex 2

Leather gloves, Precision Tex 2

10, verde/bianco, 250 mm, 0,7 - 0,9 mm
Codice catalogo 112-4111
Nome del fornitore:   HONEYWELL SAFETY
Leather gloves, Precision Tex 2
Codice catalogo 112-4111
Nome del fornitore:  HONEYWELL SAFETY
Leather gloves, Precision Tex 2
Leather gloves, Precision Tex 2

10, verde/bianco, 250 mm, 0,7 - 0,9 mm

Price
Richiedi prezzo
Log in per disponibilità
1 * 10 PAIRE
Product Details & Documents

These Gunn cut gloves made of mixed grain leather provide good mechanical resistance.

  • Gunn cut, winged thumb
  • Both supple and comfortable as a result of its jersey back and grain leather, this glove provides an excellent touch and good grip
  • Green cotton interlock back ensures better absorption of perspiration
  • Elasticated at the back, the glove offers good support

Ideal for all mechanical handlings requiring good dexterity.

Certifications: EN 388:2016 (2121X); Cat. II.

Specifiche
  • Dimensione del guanto:
    10
  • Colore:
    verde/bianco
  • Lunghezza:
    250 mm
  • Seq:
    05
  • Spessore:
    0,7 - 0,9 mm
  • Monouso/riutilizzabile:
    riutilizzabile
  • Norma EN:
    EN 388:2016 (2121X)
  • Cat. No.:
    112-4111
  • Ambidestro/mano specifico:
    mano specifico
  • Tipo polsino:
    Elastico
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.