n-Dodecil-β-D-maltoside
Bottiglia di vetro, 1 g
Catalog # PIER89902
Supplier: Thermo Fisher Scientific
n-Dodecil-β-D-maltoside
Catalog # PIER89902
Supplier: Thermo Fisher Scientific
CAS Number: 69227-93-6
Jump to:
Product Details & Documents
Synonyms: DDM, Lauryl-β-D-maltoside, Dodecyl-β-D-maltopyranoside, (2R,3R,4S,5S,6R)-2-(((2R,3S,4R,5R,6R)-6-(Dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)tetrahydro-2H-pyran-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, N-Dodecyl-Beta-Maltoside
- CAS Number:69227-93-6
- Formula:C₂₄H₄₆O₁₁
- MDL Number:MFCD00043012
- Molecular Weight:510.6 g/mol
- Storage Temperature:Congelamento
Specifications
Specifications
- Tipo di conf.:Bottiglia di vetro
- Conf:1 g



