VWR®, Dropping Bottles, PTFE
25 ml, 80 mm, 33 mm
Catalog # 216-0456
Supplier: VWR Collection
Jump to:
Product Details & Documents
In PTFE, bianco.
- Parete flessibile per un semplice utilizzo
- Tappo a vite
- Resistenza e inerzia chimica garantite
Suitable for dropping-out aggressive chemicals.
Documents
CoC VWR Dropping Bottles, PTFE.pdf
Conformity declaration
Specifications
- Ø est:33 mm
- Capacità:25 ml
- Altezza:80 mm