Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
VWR®, Crogioli, PTFE

VWR®, Crogioli, PTFE

25 ml, 44×26 mm
  216-0451
VWR®, Crogioli, PTFE
  216-0451
VWR®, Crogioli, PTFE
VWR®, Crogioli, PTFE

25 ml, 44×26 mm

 
SFr. 37.30
   
1 * 2 ST
 

Sagomati isostaticamente in PTFE puro, a norma FDA

  • Resistenza e inerzia chimica
  • Infrangibili
  • Resistenti a una temperatura massima di 280 °C
 
  • Ø×A:
    44×26 mm
  • Capacità:
    25 ml
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.