Jars with screw cap
20 ml, 38 mm, 28 mm
Catalog # SEMA5743
Supplier: SEMADENI
Jump to:
Product Details & Documents
Jars, made of PP, with tight screw cap.
Suitable for powders, granules, pastes, ointments, waxes, cosmetics and dermatology products.
Specifications
- Ø est:38 mm
- Capacità:20 ml
- Altezza:28 mm