Agarose, SeaKem® HE
Agarose, SeaKem® HE, Bottiglia di vetro
Catalog # 733-1555
Supplier: LONZA
Jump to:
Product Details & Documents
SeaKem® HE agarose is a high EEO agarose that provides enhanced resolution in immunoelectrophoresis (IEP), crossed-IEP, counterimmunoelectrophoresis (CIEP), and serum protein electrophoresis.
Specifications
- Descrizione:Agarose, SeaKem® HE
- Seq:02
- Tipo di conf.:Bottiglia di vetro