Orders LinkContactUsLinkComponent
Flasks, flat bottom with standard ground joint
500 ml, 29/32 NS
  NEUB0752-29-0500
Flasks, flat bottom with standard ground joint
  NEUB0752-29-0500
Flasks, flat bottom with standard ground joint
500 ml, 29/32 NS
SFr. 121.00
1 * 1 ST
Borosilicate glass 3,3, amber.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Amber stained glass provides protection for materials sensitive to light.
  • Cono femmina:
    29/32 NS
  • Capacità:
    500 ml
  • Seq:
    05