Labels, hazard warning
Hazard labels: foot protection, —, Poliestere, nero on giallo, 100×73 mm, 250 labels/roll
Codice catalogo BRDY250108
Nome del fornitore: Brady
Vai a
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specifiche
- Colore:nero on giallo
- Descrizione:Hazard labels: foot protection
- Dimensioni:100×73 mm
- Materiale:Poliestere
- Seq:19
- Modello:—
- Confezione:250 labels/roll