Orders LinkContactUsLinkComponent
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
10 mm, 29,2 NS
Codice catalogo SCOT286415006
Nome del fornitore:   DWK Life Sciences
Stopcocks, single-way, DURAN®
Codice catalogo SCOT286415006
Nome del fornitore:  DWK Life Sciences
Stopcocks, single-way, DURAN®
Stopcocks, single-way, DURAN®
10 mm, 29,2 NS
Price
SFr. 114.00
Log in per disponibilità
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear, with glass or PTFE keys.
  • Replaceable NS keys
  • Screw thread retaining device
Straight bore.
Specifiche
  • Ø foro:
    10 mm
  • Seq:
    32
  • Rubinetto:
    29,2 NS