Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Supelco® Organic Reference Standard, Cyclohexene

Supelco® Organic Reference Standard, Cyclohexene

Cyclohexene, Analytical standard, bottiglia in vetro, 5 mL
Catalog # 44028-5ML-F
Supplier:   Merck
Supelco® Organic Reference Standard, Cyclohexene
Catalog # 44028-5ML-F
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Cyclohexene
Supelco® Organic Reference Standard, Cyclohexene

Cyclohexene, Analytical standard, bottiglia in vetro, 5 mL

CAS Number:  
Price
SFr. 55.50
Log in for availability
1 * 5 mL
Product Details & Documents
  • GHS signal: GHS02, GHS06, GHS08

Caution: P210 - P233 - P240 - P273 - P301 + P310 - P331

Specifications

Specifications

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    5 mL
  • Standard:
    Analytical standard
  • Descrizione:
    Cyclohexene
  • Descrizione classe di pericolo:
    H225 - H302 - H304 - H411
  • N. MDL:
    MFCD00001539
  • Densità:
    0,811 g/ml at 25 °C (lit.)
  • Synonym(s):
    Tetrahydrobenzene
  • CAS No.:
    110-83-8
  • Peso molecolare:
    82,14
  • Cat. No.:
    44028-5ML-F
  • M-Clarity quality level:
    MQ100
  • Condizioni di conservazione:
    2...30 °C
  • Punto di ebollizione:
    83 °C (lit.)
  • Punto di fusione:
    –104 °C (lit.)
  • Condizioni di spedizione:
    STD
  • Punto di infiammabilità:
    –12 °C - closed cup

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.