n-Eptano, Supelco®
51730-50ML
: 142-82-5
: high purity solvents
- :142-82-5
- :H₃C(CH₂)₅CH₃
- :MFCD00009544
- :100.20 g/mol
- :13,04679
- :98,4 °C (1013 hPa)
- :–90,6 °C
- :0,6838 g/cm³ (20 °C)
- :–4 °C
- :Ambiente
- :1206
- :3,II
- :M-Clarity™ quality level = MQ100
- :205-563-8
Tipo di conf.:
bottiglia in vetro
Conf:
50 mL